missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-propylpropane-1,3-diol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-Methyl-2-propylpropane-1,3-diol
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-2-propylpropane-1,3-diol |
| CAS | 78-26-2 |
| Analyte or Component Names | 2-Methyl-2-propylpropane-1,3-diol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H16 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Methyl-2-propylpropane-1,3-diol, Carisoprodol Imp. C (EP), Carisoprodol Impurity C |
| InChI Formula | InChI=1S/C7H16O2/c1-3-4-7(2,5-8)6-9/h8-9H,3-6H2,1-2H3 |
| SMILES | CCCC(C)(CO)CO |
| IUPAC Name | 2-methyl-2-propylpropane-1,3-diol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction