Learn More
2-Methoxynicotinic acid, 98%
Product Code. 11464604
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11464604
Brand: Thermo Scientific Alfa Aesar A13193.03
Request Bulk or Custom Format
CAS: 16498-81-0 | C7H6NO3 | 152.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16498-81-0
Molecular Formula C7H6NO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00085110
InChI Key FTEZJSXSARPZHJ-UHFFFAOYSA-M
Synonym 2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh
PubChem CID 708093
IUPAC Name 2-methoxypyridine-3-carboxylic acid
SMILES COC1=NC=CC=C1C([O-])=O
Specifications
Melting Point 147°C to 151°C
Quantity 1 g
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 2-Methoxynicotinic acid