accessibility menu, dialog, popup

UserName

CAS RN 16498-81-0

H3CONOO

CAS RN 16498-81-0

IUPAC Name: 2-methoxypyridine-3-carboxylate
Synonyms: 2-methoxypyridine-3-carboxylate
Molecular Weight (g/mol): 152.13
Molecular Formula: C7H6NO3
InChi Key: FTEZJSXSARPZHJ-UHFFFAOYSA-M
SMILES: COC1=NC=CC=C1C([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)