missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy Estrone, TRC

Product Code. 30009925
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009925 50 mg 50mg
30007329 25 mg 25mg
30009052 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009925 Supplier Toronto Research Chemicals Supplier No. TRCM26252050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362-08-3
Molecular Formula C19 H24 O3
Molecular Weight (g/mol) 300.39
Synonym 2-Methoxy Estrone, 3-Hydroxy-2-methoxyestra-1,3,5(10)-trien-17-one, 2-Hydroxyestrone 2-Methyl Ether, 2-Methoxy-3-hydroxyestra-1,3,5(10)-trien-17-one, 2-Methoxy-17-oxoestra-1,3,5(10)-trien-3-ol, 3-Hydroxy-2-methoxyestra-1,3,5(10)-trien-17-one, GSS 33, 2-Methoxyestrone;
IUPAC Name (8R,9S,13S,14S)-3-hydroxy-2-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
SMILES COc1cc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc2cc1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H24O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,20H,3-8H2,1-2H3/t12-,13+,15-,19-/m0/s1
Formula Weight 300.1725
Percent Purity >95%
Chemical Name or Material 2-Methoxyestrone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.