CAS RN 362-08-3
CAS RN 362-08-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methoxy Estrone, TRC
CAS: 362-08-3 Molecular Formula: C19 H24 O3 Molecular Weight (g/mol): 300.39 Synonym: 2-Methoxy Estrone,3-Hydroxy-2-methoxyestra-1,3,5(10)-trien-17-one,2-Hydroxyestrone 2-Methyl Ether,2-Methoxy-3-hydroxyestra-1,3,5(10)-trien-17-one,2-Methoxy-17-oxoestra-1,3,5(10)-trien-3-ol,3-Hydroxy-2-methoxyestra-1,3,5(10)-trien-17-one,GSS 33,2-Methoxyestrone; IUPAC Name: (8R,9S,13S,14S)-3-hydroxy-2-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one SMILES: COc1cc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc2cc1O