Learn More
2-Isobutylthiazole, 99%
Product Code. 11434804
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11434804
Brand: Thermo Scientific Alfa Aesar A13759.06
Request Bulk or Custom Format
CAS: 18640-74-9 | C7H11NS | 141.232 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18640-74-9
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.232
MDL Number MFCD00005334
InChI Key CMPVUVUNJQERIT-UHFFFAOYSA-N
Synonym 2-isobutylthiazole, 2-isobutyl-1,3-thiazole, 2-2-methylpropyl thiazole, thiazole, 2-isobutyl, 2-2-methylpropyl-1,3-thiazole, 2-isobutyl thiazole, thiazole, 2-2-methylpropyl, iso-butyl thiazole, fema no. 3134
PubChem CID 62725
IUPAC Name 2-(2-methylpropyl)-1,3-thiazole
SMILES CC(C)CC1=NC=CS1
Specifications
Density 0.998
Boiling Point 172°C to 173°C
Flash Point 58°C (136°F)
Odor Characteristic
Refractive Index 1.496
Quantity 5 g
UN Number UN1993
Beilstein 507823
Formula Weight 141.24
Percent Purity 99%
Chemical Name or Material 2-Isobutylthiazole