CAS RN 18640-74-9
CAS RN 18640-74-9
IUPAC Name:
2-(2-methylpropyl)-1,3-thiazole
Synonyms:
isobutylthiazole
Molecular Weight (g/mol):
141.23
Molecular Formula:
C7H11NS
InChi Key:
CMPVUVUNJQERIT-UHFFFAOYSA-N
SMILES:
CC(C)CC1=NC=CS1
1
–
2
of
2
results
1
2-Isobutylthiazole, 99%
CAS: 18640-74-9 Molecular Formula: C7H11NS Molecular Weight (g/mol): 141.232 MDL Number: MFCD00005334 InChI Key: CMPVUVUNJQERIT-UHFFFAOYSA-N Synonym: 2-isobutylthiazole,2-isobutyl-1,3-thiazole,2-2-methylpropyl thiazole,thiazole, 2-isobutyl,2-2-methylpropyl-1,3-thiazole,2-isobutyl thiazole,thiazole, 2-2-methylpropyl,iso-butyl thiazole,fema no. 3134 PubChem CID: 62725 IUPAC Name: 2-(2-methylpropyl)-1,3-thiazole SMILES: CC(C)CC1=NC=CS1
2
2-Isobutylthiazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.