Learn More
2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals
Product Code. 15408377
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408377 5 g 5g
15498367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15408377 Supplier Thermo Scientific Alfa Aesar Supplier No. H51875.06
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
CAS: 4102-54-9 | Molecular Formula: C8H10IN | Molecular Weight: 247.079 g/mol
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 4102-54-9
Molecular Formula C8H10IN
Molecular Weight (g/mol) 247.079
MDL Number MFCD07779005
InChI Key ODPOIEACUVQCBZ-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-iodoaniline, 2-iodo-4,6-di-methylaniline, benzenamine,2-iodo-4,6-dimethyl
PubChem CID 14040290
IUPAC Name 2-iodo-4,6-dimethylaniline
SMILES CC1=CC(=C(C(=C1)I)N)C
Spezifikation
Melting Point 64°C to 65°C
Quantity 5 g
UN Number UN2811
Formula Weight 247.08
Percent Purity 98%
Chemical Name or Material 2-Iodo-4,6-dimethylaniline