accessibility menu, dialog, popup

UserName

CAS RN 4102-54-9

CH3CH3NH2I

CAS RN 4102-54-9

IUPAC Name: 2-iodo-4,6-dimethylaniline
Synonyms: 2-iodo-4,6-dimethylaniline
Molecular Weight (g/mol): 247.08
Molecular Formula: C8H10IN
InChi Key: ODPOIEACUVQCBZ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N)C(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results