missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam 2-Heptyl-3-hydroxy-4-quinolone, Intercellular Signal Compound, Solid, 25 mg
Description
- Molecular weight: 259.34 g/mol
- Purity: >96%
- Form: Solid
- Composition: 100% 2-Heptyl-3-hydroxy-4(1H)-quinolinone
- Function: Quorum sensing signaling molecule used to induce and study the regulation of virulence genes
- Storage: Store at ambient temperature
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% 2-Heptyl-3-hydroxy-4(1H)-quinolinone |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | 2-Heptyl-3-hydroxy-4-quinolone |
| Quantity | 25 mg |
| For Use With (Application) | Quorum sensing, virulence gene regulation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?