missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15422256
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15422256 250 mg 250mg
15412256 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15422256

Brand: Thermo Scientific Alfa Aesar H55727.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874289-23-3
Molecular Formula C8H9BFNO3
Molecular Weight (g/mol) 196.972
MDL Number MFCD11856043
InChI Key STYRVOIKGOQNAD-UHFFFAOYSA-N
Synonym 2-fluoro-4-methylcarbamoyl phenyl boronic acid, 2-fluoro-4-methylcarbamoyl benzeneboronic acid, 4-n-methylaminocarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methylcarbamoyl phenylboronic acid
PubChem CID 57497292
IUPAC Name [2-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O

Specifications

Quantity 250 mg
Formula Weight 196.97
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.