accessibility menu, dialog, popup

UserName

CAS RN 874289-23-3

BOHNCH3FHOOH

CAS RN 874289-23-3

IUPAC Name: [2-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 2-fluoro-4-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 196.97
Molecular Formula: C8H9BFNO3
InChi Key: STYRVOIKGOQNAD-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC(F)=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results