missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxyphenylacetic acid, 97%, Thermo Scientific™

Product Code. 15454447
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454447 250 mg 250mg
15444447 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454447 Supplier Thermo Scientific Alfa Aesar Supplier No. H31810.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 883531-28-0
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.166
MDL Number MFCD06800744
InChI Key WGODSCYEKWDSAV-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxyphenylacetic acid, 2-2-fluoro-4-methoxyphenyl acetic acid, 2-fluoro-4-methoxyphenyl acetic acid, 2-fluoro-4-methoxylphenylacetic acid, 2-fluoro-4-methoxyphenylaceticacid, pubchem4172, 2-fluoro-4-methoxy acid, ksc495c2t, 4-carboxymethyl-3-fluoroanisole, 2-fluoro-4-methoxyphenylacetic acid, jrd
PubChem CID 19361978
IUPAC Name 2-(2-fluoro-4-methoxyphenyl)acetic acid
SMILES COC1=CC(=C(C=C1)CC(=O)O)F

Specifications

Quantity 250 mg
Formula Weight 184.17
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methoxyphenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.