accessibility menu, dialog, popup

UserName

CAS RN 883531-28-0

H3COOHOF

CAS RN 883531-28-0

IUPAC Name: 2-(2-fluoro-4-methoxyphenyl)acetic acid
Synonyms: (2-fluoro-4-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: WGODSCYEKWDSAV-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(CC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results