missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxybenzoyl chloride, 95%, Thermo Scientific™

Product Code. 15404257
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404257 1 g 1g
15414257 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404257 Supplier Thermo Scientific Alfa Aesar Supplier No. H31644.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 321-24-4
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD08146649
InChI Key UHLDNUXKNREILG-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxybenzoylchloride, benzoyl chloride, 2-fluoro-4-methoxy, pubchem4965, benzoyl chloride,2-fluoro-4-methoxy
PubChem CID 14484714
IUPAC Name 2-fluoro-4-methoxybenzoyl chloride
SMILES COC1=CC(=C(C=C1)C(=O)Cl)F

Specifikationer

Melting Point 40°C to 42°C
Odor Pungent
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 188.59
Percent Purity 95%
Chemical Name or Material 2-Fluoro-4-methoxybenzoyl chloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.