accessibility menu, dialog, popup

UserName

H3COOClF

CAS RN 321-24-4

IUPAC Name: 2-fluoro-4-methoxybenzoyl chloride
Synonyms: 2-fluoro-4-methoxybenzoyl chloride
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: UHLDNUXKNREILG-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results