missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxybenzonitrile, 97%, Thermo Scientific™

Product Code. p-7031449
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15405267

Brand: Thermo Scientific Alfa Aesar H32531.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-4-methoxybenzonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94610-82-9
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00509374
InChI Key HWKUZTFIZATJPM-UHFFFAOYSA-N
Synonym 3-fluoro-4-cyanoanisole, 2-fluoro-4-methoxy-benzonitrile, benzonitrile, 2-fluoro-4-methoxy, 3-fluoro-4-cyano anisole, 4-cyano-3-fluoroanisole, buttpark 81\01-98, 2-fluoro-4-methoxybenzenecarbonitrile, pubchem4783, 2-fluoro-4-methoxybe
PubChem CID 639878
IUPAC Name 2-fluoro-4-methoxybenzonitrile
SMILES COC1=CC(F)=C(C=C1)C#N

Specifications

Melting Point 62°C to 65°C
Quantity 1 g
UN Number UN3439
Formula Weight 151.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methoxybenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.