accessibility menu, dialog, popup

UserName

CAS RN 94610-82-9

H3CONF

CAS RN 94610-82-9

IUPAC Name: 2-fluoro-4-methoxybenzonitrile
Synonyms: 2-fluoro-4-methoxybenzonitrile
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: HWKUZTFIZATJPM-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results