Learn More
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
Product Code. 15438045
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438045 5 g 5g
15428045 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15438045 Supplier Thermo Scientific Alfa Aesar Supplier No. H32292.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride, is used as a pharmaceutical intermediate.
Solubility
It reacts with water.
Notes
Moisture&water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017778-34-5
Molecular Formula C9H2ClF7O
Molecular Weight (g/mol) 294.553
MDL Number MFCD09258670
InChI Key RBJVFNYOFYHETI-UHFFFAOYSA-N
Synonym 2-fluoro-4,6-bis trifluoromethyl benzoyl chloride
PubChem CID 46737582
IUPAC Name 2-fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
SMILES C1=C(C=C(C(=C1C(F)(F)F)C(=O)Cl)F)C(F)(F)F
Specifications
Refractive Index 1.4146
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 294.55
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride