Learn More
2-Ethylhexyl 4-dimethylaminobenzoate, 99%
Product Code. 15400518
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 15400518
Brand: Thermo Scientific Alfa Aesar H54136.AE
Request Bulk or Custom Format
CAS: 21245-02-3 | C17H27NO2 | 277.408 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21245-02-3
Molecular Formula C17H27NO2
Molecular Weight (g/mol) 277.408
MDL Number MFCD00017526
InChI Key WYWZRNAHINYAEF-UHFFFAOYSA-N
Synonym 2-ethylhexyl 4-dimethylamino benzoate, padimate o, escalol 507, padimate-o, octyl dimethyl paba, arlatone uvb, benzoic acid, 4-dimethylamino-, 2-ethylhexyl ester, 2-ethylhexyl p-dimethylaminobenzoate, eusolex 6007, sundown
PubChem CID 30541
IUPAC Name 2-ethylhexyl 4-(dimethylamino)benzoate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C
Specifications
Density 0.995
Boiling Point 325°C
Flash Point 113°C (235°F)
Refractive Index 1.542
Quantity 100 mL
Merck Index 14,3232
Formula Weight 277.4
Percent Purity 99%
Chemical Name or Material 2-Ethylhexyl 4-dimethylaminobenzoate