accessibility menu, dialog, popup

UserName

CAS RN 21245-02-3

H3CCH3OONCH3CH3

CAS RN 21245-02-3

IUPAC Name: 2-ethylhexyl 4-(dimethylamino)benzoate
Synonyms: padimate O octyl dimethyl p-aminobenzoate padimate-O padimate-o arlatone 507 ethylhexyl dimethyl paba octyl dimethyl paba octyl dimethyl aminobenzoate escalol 507 padimate-o preparation
Molecular Weight (g/mol): 277.41
Molecular Formula: C17H27NO2
InChi Key: WYWZRNAHINYAEF-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results