missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1H-indene, 97%

Product Code. 11496306
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11496306

Brand: Thermo Scientific Alfa Aesar B23104.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 17059-50-6 | C11H12 | 144.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17059-50-6
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD01321239
InChI Key BSHJHVHMLRKHBZ-UHFFFAOYSA-N
PubChem CID 582328
IUPAC Name 2-ethyl-1H-indene
SMILES CCC1=CC2=CC=CC=C2C1

Specifications

Density 0.964
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point 87°C (190°F)
Refractive Index 1.559
Quantity 5 g
Synonym 2-ethylindene, 2-ethyl-1-h-indene, 1h-indene, 2-ethyl, pubchem5396, 1h-indene,2-ethyl, acmc-20a5x3
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 2-Ethyl-1H-indene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.