accessibility menu, dialog, popup

UserName

CAS RN 17059-50-6

H3C

CAS RN 17059-50-6

IUPAC Name: 2-ethyl-1H-indene
Synonyms: 2-ethyl-1H-indene
Molecular Weight (g/mol): 144.22
Molecular Formula: C11H12
InChi Key: BSHJHVHMLRKHBZ-UHFFFAOYSA-N
SMILES: CCC1=CC2=CC=CC=C2C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results