missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxynicotinic acid, 98%, Thermo Scientific™

Product Code. 11499916
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11499916

Brand: Thermo Scientific Alfa Aesar B20887.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35969-54-1
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00128788
InChI Key XCMJQQOMGWGGSI-UHFFFAOYSA-N
Synonym 2-ethoxynicotinic acid, 2-ethoxy-nicotinic acid, 3-pyridinecarboxylic acid, 2-ethoxy, 3-pyridinecarboxylicacid, 2-ethoxy, pubchem16563, acmc-1aft2, 2-ethoxy-3-pyridinecarboxylic acid, 2-ethoxypyridine-3-carboxylic acid
PubChem CID 2736345
IUPAC Name 2-ethoxypyridine-3-carboxylic acid
SMILES CCOC1=C(C=CC=N1)C(=O)O

Specifications

Quantity 50 g
Formula Weight 167.16
Percent Purity 98%
Chemical Name or Material 2-Ethoxynicotinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.