accessibility menu, dialog, popup

UserName

CAS RN 35969-54-1

CH3ONOO

CAS RN 35969-54-1

IUPAC Name: 2-ethoxypyridine-3-carboxylate
Synonyms: 2-ethoxypyridine-3-carboxylate
Molecular Weight (g/mol): 166.16
Molecular Formula: C8H8NO3
InChi Key: XCMJQQOMGWGGSI-UHFFFAOYSA-M
SMILES: CCOC1=NC=CC=C1C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results