missing translation for 'onlineSavingsMsg'
Learn More

Abcam 2'-Deoxyuridine-5'-monophosphate Disodium Salt, Solid, >98% Purity, 1 g

Product Code. 32116915 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116915 Supplier Abcam Supplier No. ab1462241g
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reference substrate for studies of dUMP analogs in potential chemotherapy applications.

  • Molecular Weight: 352.1 g/mol
  • Purity: >98%
  • Applications: Reference substrate used by thymidylate synthetase (EC 2.1.1.45) for the de novo production of dTMP; used in studies of dUMP analogs for potential chemotherapy applications
  • Form: Solid
  • Storage Buffer: 100% 2'-Deoxy-5'-uridylic acid disodium salt

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% 2'-Deoxy-5'-uridylic acid disodium salt
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate dUMP
Target Molecule Thymidylate Synthetase
Formulation 100% 2'-Deoxy-5'-uridylic acid disodium salt
Quantity 1 g
For Use With (Application) Thymidylate synthetase enzyme studies, dUMP analog research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.