missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam 2'-Deoxycytidine 5'-Monophosphate, UMP/CMP Kinase Substrate, 250 mg
Description
- Molecular Weight: 307.2 g/mol
- Function: Nucleoside monophosphate used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP, which supports DNA biosynthesis upon phosphorylation to dCTP
- Composition: 100% 2'-Deoxycytidine 5'-monophosphate free acid
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | 2'-Deoxycytidine 5'-monophosphate |
| Target Molecule | 2'-Deoxycytidine 5'-monophosphate |
| Quantity | 250 mg |
| For Use With (Application) | DNA biosynthesis, UMP/CMP kinase substrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?