Learn More
2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%
Product Code. 15416526
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416526 250 mg 250mg
15406526 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15416526 Supplier Thermo Scientific Alfa Aesar Supplier No. H61451.MD
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
CAS: 1218790-84-1 | C8H5BF3NO2 | 214.938 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1218790-84-1
Molecular Formula C8H5BF3NO2
Molecular Weight (g/mol) 214.938
InChI Key DELAZGQTKSRLPA-UHFFFAOYSA-N
Synonym 2-cyano-4-trifluoromethyl phenylboronic acid, 2-cyano-4-trifluoromethyl phenyl boronic acid, acmc-209ail, 2-cyano-4-trifluoromethyl phenyl boronicacid
PubChem CID 53216599
IUPAC Name [2-cyano-4-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(F)(F)F)C#N)(O)O
Specifications
Quantity 250 mg
Formula Weight 214.94
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Cyano-4-(trifluoromethyl)benzeneboronic acid