missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 11314426
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314426 250 mg 250mg
11304426 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314426 Supplier Thermo Scientific Alfa Aesar Supplier No. L20024.MD
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628691-93-0 | Molecular Formula: C6H3ClFNO2 | Molecular Weight: 175.543 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628691-93-0
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.543
MDL Number MFCD04038325
InChI Key NWNWBLRKHOVSEL-UHFFFAOYSA-N
Synonym 2-chloro-3-fluoroisonicotinic acid, 2-chloro-3-fluoro-4-carboxypyridine, 4-pyridinecarboxylic acid, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine-4-carboxylic acid, 2-chloro-3-fluoro-isonicotinic acid, 2-chloro-3-fluoro-4-pyridinecarboxylic acid, pubchem17055, 2-chloro-3-fluoroisonicotinicacid, acmc-1bfbj, g00017-watson-int
PubChem CID 2782812
IUPAC Name 2-chloro-3-fluoropyridine-4-carboxylic acid
SMILES C1=CN=C(C(=C1C(=O)O)F)Cl

Specifications

Melting Point ∼200°C
Quantity 250 mg
Formula Weight 175.55
Percent Purity 95%
Chemical Name or Material 2-Chloro-3-fluoropyridine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.