accessibility menu, dialog, popup

UserName

CAS RN 628691-93-0

IUPAC Name: 2-chloro-3-fluoropyridine-4-carboxylic acid
Synonyms: 2-chloro-3-fluoropyridine-4-carboxylic acid
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: NWNWBLRKHOVSEL-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results