missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)benzaldehyde, 97%

Product Code. p-7028705
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443387 250 mg 250mg
11342089 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443387 Supplier Thermo Scientific Alfa Aesar Supplier No. H26109.MD
Add to basket
This item is not returnable. View return policy

CAS: 102684-91-3 | C8H4BrF3O | 253.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102684-91-3
Molecular Formula C8H4BrF3O
Molecular Weight (g/mol) 253.02
MDL Number MFCD04973760
InChI Key CSOBJYGHQOLWOD-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl benzaldehyde, 4-bromo-3-formylbenzotrifluoride, 2-bromo-5-1,1-dimethylethyl-benzaldehyde, benzaldehyde, 2-bromo-5-1,1-dimethylethyl, benzaldehyde, 2-bromo-5-trifluoromethyl, paragos 530289, pubchem1428, pubchem4179, ksc498c1d, bromo 5-trifluoromethyl benzaldehyde
PubChem CID 7018047
IUPAC Name 2-bromo-5-(trifluoromethyl)benzaldehyde
SMILES FC(F)(F)C1=CC(C=O)=C(Br)C=C1

Specifications

Refractive Index 1.511
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 253.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.