Learn More
2-Bromo-5-fluorobenzotrifluoride, 97%, Thermo Scientific™
Product Code. 11476906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476906 25 g 25g
11456906 1 g 1g
11466906 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11476906 Supplier Thermo Scientific Alfa Aesar Supplier No. B24005.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40161-55-5
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD00040937
InChI Key AIDVAZGOACECLJ-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzotrifluoride, 1-bromo-4-fluoro-2-trifluoromethyl benzene, 1-bromo-2-trifluoromethyl-4-fluorobenzene, 4-fluoro-2-trifluoromethyl bromobenzene, 2-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, benzene, 1-bromo-4-fluoro-2-trifluoromethyl, 2-bromo-5-fluoro benzotrifluoride, pubchem1637, pubchem1643
PubChem CID 520983
IUPAC Name 1-bromo-4-fluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC(=C(Br)C=C1)C(F)(F)F
Specifications
Density 1.72
Boiling Point 136°C
Flash Point 63°C (145°F)
Refractive Index 1.465
Quantity 25 g
Beilstein 2643544
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-fluorobenzotrifluoride