Learn More
2-Amino-6-mercaptopurine-9-D-riboside Hydrate, TRC
Product Code. 30005999
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005999 2.5 g 2.5g
30006915 1 g 1g
17931165 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005999 Supplier Toronto Research Chemicals Supplier No. TRCA6118502.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 345909-25-3
Molecular Formula C10 H13 N5 O4 S . x(H2 O)
Molecular Weight (g/mol) 299.31
Synonym Thioguanosine, NSC-29422;
IUPAC Name (2R,3R,4S,5R)-2-(2-amino-6-sulfanyl-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol;hydrate
SMILES O.Nc1nc(S)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c2n1
Specifications
Analyte or Component Names 2-Amino-6-mercaptopurine-9-D-riboside Hydrate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H13N5O4S.H2O/c11-10-13-7-4(8(20)14-10)12-2-15(7)9-6(18)5(17)3(1-16)19-9;/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,20);1H2/t3-,5-,6-,9-;/m1./s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 2.5 g
Chemical Name or Material 2-Amino-6-mercaptopurine-9-D-riboside Hydrate