Learn More
2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%
Product Code. 11327595
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327595 5 g 5g
11317595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11327595 Supplier Thermo Scientific Alfa Aesar Supplier No. L05778.06
Request Bulk or Custom Format
CAS: 57235-50-4 | C5H7N3S | 141.19 g/mol
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.
Solubility
Soluble in Methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at -20°C.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57235-50-4
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.19
MDL Number MFCD00051649
InChI Key AVLUMBXGKFNNAS-UHFFFAOYSA-N
Synonym 2-amino-5-cyclopropyl-1,3,4-thiadiazole, 5-cyclopropyl 1,3,4 thiadiazol-2-ylamine, 5-cyclopropyl-1,3,4-thiadiazole-2-ylamine, acmc-20ao4n, enamine_005503, 5-cyclopropyl-2-amino-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine,5-cyclopropyl, 5-cyclopropyl-1,3,4 thiadiazol-2-ylamine
PubChem CID 717021
IUPAC Name 5-cyclopropyl-1,3,4-thiadiazol-2-amine
SMILES NC1=NN=C(S1)C1CC1
Specifications
Melting Point 212°C to 216°C
Quantity 5 g
Beilstein 124356
Solubility Information Soluble in Methanol. Insoluble in water.
Formula Weight 141.2
Percent Purity 98%
Chemical Name or Material 2-Amino-5-cyclopropyl-1,3,4-thiadiazole