CAS RN 57235-50-4
CAS RN 57235-50-4
IUPAC Name:
5-cyclopropyl-1,3,4-thiadiazol-2-amine
Synonyms:
5-cyclopropyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol):
141.19
Molecular Formula:
C5H7N3S
InChi Key:
AVLUMBXGKFNNAS-UHFFFAOYSA-N
SMILES:
NC1=NN=C(S1)C1CC1
1
–
2
of
2
results
1
2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%
CAS: 57235-50-4 Molecular Formula: C5H7N3S Molecular Weight (g/mol): 141.19 MDL Number: MFCD00051649 InChI Key: AVLUMBXGKFNNAS-UHFFFAOYSA-N Synonym: 2-amino-5-cyclopropyl-1,3,4-thiadiazole,5-cyclopropyl 1,3,4 thiadiazol-2-ylamine,5-cyclopropyl-1,3,4-thiadiazole-2-ylamine,acmc-20ao4n,enamine_005503,5-cyclopropyl-2-amino-1,3,4-thiadiazole,1,3,4-thiadiazol-2-amine,5-cyclopropyl,5-cyclopropyl-1,3,4 thiadiazol-2-ylamine PubChem CID: 717021 IUPAC Name: 5-cyclopropyl-1,3,4-thiadiazol-2-amine SMILES: NC1=NN=C(S1)C1CC1
2
2-Amino-5-cyclopropyl-1,3,4-thiadiazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.