missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-hydroxy-2-(hydroxymethyl)propyl Phosphate Barium Salt, TRC

Product Code. 30133916
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133916 100 mg 100mg
30004822 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133916 Supplier Toronto Research Chemicals Supplier No. TRCA611560100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114252-50-5
Molecular Formula C4 H10 N O6 P . Ba
Molecular Weight (g/mol) 336.43
Synonym 2-Amino-2-(hydroxymethyl)-,3-propanediol Phosphate Barium Salt, Trometamol Phosphoric Acid Barium Salt, Fosfomycin EP Impurity C Barium Salt, Trometamine Phosphate Barium Salt, Fosfomycin EP Impurity C (Barium Salt)
IUPAC Name [2-amino-3-hydroxy-2-(hydroxymethyl)propyl] phosphate;barium(2+)
SMILES [Ba+2].NC(CO)(CO)COP(=O)([O-])[O-]

Specifications

Quantity 100 mg
Shipping Condition Room Temperature
InChI Formula InChI=1S/C4H12NO6P.Ba/c5-4(1-6,2-7)3-11-12(8,9)10;/h6-7H,1-3,5H2,(H2,8,9,10);/q;+2/p-2
Formula Weight 336.9298
Percent Purity >95%
Chemical Name or Material 2-Amino-3-hydroxy-2-(hydroxymethyl)propyl phosphate barium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.