CAS RN 114252-50-5
CAS RN 114252-50-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-3-hydroxy-2-(hydroxymethyl)propyl Phosphate Barium Salt, TRC
CAS: 114252-50-5 Molecular Formula: C4 H10 N O6 P . Ba Molecular Weight (g/mol): 336.43 Synonym: 2-Amino-2-(hydroxymethyl)-,3-propanediol Phosphate Barium Salt,Trometamol Phosphoric Acid Barium Salt,Fosfomycin EP Impurity C Barium Salt,Trometamine Phosphate Barium Salt,Fosfomycin EP Impurity C (Barium Salt) IUPAC Name: [2-amino-3-hydroxy-2-(hydroxymethyl)propyl] phosphate;barium(2+) SMILES: [Ba+2].NC(CO)(CO)COP(=O)([O-])[O-]