Learn More
2-Amino-3-ethyl-6-methylpyridine, 97%
Product Code. 11499346
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499346 5 g 5g
11409356 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11499346 Supplier Thermo Scientific Alfa Aesar Supplier No. A19048.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 41995-31-7 | C8H12N2 | 136.198 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41995-31-7
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00130077
InChI Key OQNDNGDJFBXUID-UHFFFAOYSA-N
Synonym 2-amino-3-ethyl-6-methylpyridine, 3-ethyl-6-methyl-2-pyridinamine, 3-ethyl-6-methyl-2-pyridylamine, acmc-1aoyl, 2-pyridinamine,3-ethyl-6-methyl, 3-ethyl-6-methyl pyridin-2-yl amine, 3-ethyl-6-methyl-pyridin-2-ylamine
PubChem CID 3016311
IUPAC Name 3-ethyl-6-methylpyridin-2-amine
SMILES CCC1=C(N=C(C=C1)C)N
Specifications
Melting Point 45°C to 48°C
Quantity 5 g
UN Number UN2811
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material 2-Amino-3-ethyl-6-methylpyridine