CAS RN 41995-31-7
CAS RN 41995-31-7
IUPAC Name:
2-amino-3-ethyl-6-methylpyridin-1-ium
Synonyms:
2-amino-3-ethyl-6-methylpyridin-1-ium
Molecular Weight (g/mol):
137.21
Molecular Formula:
C8H13N2
InChi Key:
OQNDNGDJFBXUID-UHFFFAOYSA-O
SMILES:
CCC1=CC=C(C)[NH+]=C1N
1
–
1
of
1
results
1
2-Amino-3-ethyl-6-methylpyridine, 97%
CAS: 41995-31-7 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.198 MDL Number: MFCD00130077 InChI Key: OQNDNGDJFBXUID-UHFFFAOYSA-N Synonym: 2-amino-3-ethyl-6-methylpyridine,3-ethyl-6-methyl-2-pyridinamine,3-ethyl-6-methyl-2-pyridylamine,acmc-1aoyl,2-pyridinamine,3-ethyl-6-methyl,3-ethyl-6-methyl pyridin-2-yl amine,3-ethyl-6-methyl-pyridin-2-ylamine PubChem CID: 3016311 IUPAC Name: 3-ethyl-6-methylpyridin-2-amine SMILES: CCC1=C(N=C(C=C1)C)N