Learn More
2-Amino-2'-O-methyladenosine, 99%, Thermo Scientific™
Product Code. 15426849
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426849 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15426849 Supplier Thermo Scientific Alfa Aesar Supplier No. J65334.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 80791-87-3
Molecular Formula C11H16N6O4
Molecular Weight (g/mol) 296.29
MDL Number MFCD01630959
InChI Key JLWUWXCKSOIFPS-YPLCUDRINA-N
Synonym 2-amino-2'-o-methyladenosine, 2'-o-methyl-2,6-diaminopurine-riboside, 2-amino-2'-o-methyl-adenosine, 2r,3r,4r,5r-5-2,6-diaminopurin-9-yl-2-hydroxymethyl-4-methoxyoxolan-3-ol, 2'-o-methyl-2-aminoadenosine, 2-nh2-2'-o-me-a, adenosine, 2-amino-2'-o-methyl, 2?-o-methyl-2,6-diaminopurine-riboside, 2,6-diamino-9-2-o-methyl-beta-d-ribofuranosyl purine, 2,6-diamino-9-2'-o-methyl-beta-d-ribofuranosyl purine
PubChem CID 9900805
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=C(N)N=C12
Specifications
Color White
Quantity 1 g
IUPAC Name (2R,3R,4R,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Formula Weight 296.28
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Amino-2'-O-methyladenosine