missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethoxynaphthalene, 98%

Product Code. 11377549
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377549 5 g 5g
11387549 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377549 Supplier Thermo Scientific Alfa Aesar Supplier No. H27971.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3469-26-9 | C12H12O2 | 188.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3469-26-9
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00004064
InChI Key PPKHAIRFQKFMLE-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dimethoxy, acmc-209i9n, ksc225e3r, 2,7-dimethoxynaphthalene, ppkhairfqkfmle-uhfffaoysa, 2,7-dimethoxy naphthalene, benzocyclobuten-1 2h-one; bicyclo 4.2.0 octa-1,3,5-trien-7-one
PubChem CID 77021
IUPAC Name 2,7-dimethoxynaphthalene
SMILES COC1=CC2=C(C=C1)C=CC(=C2)OC

Specifications

Melting Point 137°C to 140°C
Quantity 5 g
Formula Weight 188.23
Percent Purity 98%
Chemical Name or Material 2,7-Dimethoxynaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.