accessibility menu, dialog, popup

UserName

CAS RN 3469-26-9

IUPAC Name: 2,7-dimethoxynaphthalene
Synonyms: 2,7-dimethoxynaphthalene
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: PPKHAIRFQKFMLE-UHFFFAOYSA-N
SMILES: COC1=CC2=CC(OC)=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results