missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxypyridine-3,5-diamine Dihydrochloride (>90%), TRC

Product Code. 30373469
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373469 100 mg
30383829 500 mg
30382443 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373469 Supplier Toronto Research Chemicals Supplier No. TRCD562203100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56216-28-5
Molecular Formula C7H11N3O2 . 2HCl
Molecular Weight (g/mol) 205.64
SMILES Cl.Cl.COc1nc(OC)c(N)cc1N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H11N3O2.ClH/c1-11-6-4(8)3-5(9)7(10-6)12-2;/h3H,8-9H2,1-2H3;1H
Formula Weight 241.0385
Chemical Name or Material 2,6-Dimethoxypyridine-3,5-diamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.