CAS RN 56216-28-5
CAS RN 56216-28-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dimethoxypyridine-3,5-diamine Dihydrochloride (>90%), TRC
CAS: 56216-28-5 Molecular Formula: C7H11N3O2 . 2HCl Molecular Weight (g/mol): 205.64 SMILES: Cl.Cl.COc1nc(OC)c(N)cc1N