Learn More
2,6-Difluorobenzyl alcohol, 97%, Thermo Scientific™
Product Code. p-7024121
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403907 5 g 5g
11413907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11403907 Supplier Thermo Scientific Alfa Aesar Supplier No. A18336.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,6-Difluorobenzyl alcohol is used to get 2-(bromomethyl)-1,3-difluorobenzene.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19064-18-7
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00004603
InChI Key LVICICZQETYOGS-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl alcohol, 2,6-difluorophenyl methanol, benzenemethanol, 2,6-difluoro, 2,6-difluorobenzylalcohol, 2,6-difluorobenzenemethanol, 2,6-difluoro-phenyl-methanol, 2,6-difluoro benzyl alcohol, 2,6-difluorophenyl methan-1-ol, pubchem4914, acmc-209etz
PubChem CID 87921
IUPAC Name (2,6-difluorophenyl)methanol
SMILES C1=CC(=C(C(=C1)F)CO)F
Specifications
Density 1.3
Boiling Point 87°C to 88°C (14 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.495
Quantity 5 g
Beilstein 2501536
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 2,6-Difluorobenzyl alcohol