accessibility menu, dialog, popup

UserName

HOFF

CAS RN 19064-18-7

IUPAC Name: (2,6-difluorophenyl)methanol
Synonyms: benzenemethanol, 2,6-difluoro-
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: LVICICZQETYOGS-UHFFFAOYSA-N
SMILES: OCC1=C(F)C=CC=C1F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)