Learn More
2,6-Dichloro-5-fluoronicotinic acid, 96%, Thermo Scientific™
Product Code. 11368766
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11368766
Brand: Thermo Scientific Alfa Aesar L19454.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82671-06-5
Molecular Formula C6H2Cl2FNO2
Molecular Weight (g/mol) 209.985
MDL Number MFCD00799517
InChI Key LTDGKGCHRNNCAC-UHFFFAOYSA-N
Synonym 2,6-dichloro-5-fluoronicotinic acid, 2,6-dichloro-5-fluoro-nicotinic acid, 2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro-pyridine-3-carboxylic acid, 2,6-dichloro-5-fluoro nicotinic acid, 3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro, pubchem10943, acmc-1bluq, ksc448a5j, 2,6-dichloro-5-fluoro-nicotinicacid
PubChem CID 2733659
IUPAC Name 2,6-dichloro-5-fluoropyridine-3-carboxylic acid
SMILES C1=C(C(=NC(=C1F)Cl)Cl)C(=O)O
Specifications
Melting Point 152°C to 155°C
Quantity 25 g
Beilstein 4311035
Formula Weight 209.99
Percent Purity 96%
Chemical Name or Material 2,6-Dichloro-5-fluoronicotinic acid