CAS RN 82671-06-5
CAS RN 82671-06-5
IUPAC Name:
2,6-dichloro-5-fluoropyridine-3-carboxylate
Synonyms:
2,6-dichloro-5-fluoropyridine-3-carboxylate
Molecular Weight (g/mol):
208.98
Molecular Formula:
C6HCl2FNO2
InChi Key:
LTDGKGCHRNNCAC-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC(F)=C(Cl)N=C1Cl
1
–
2
of
2
results
2
2,6-Dichloro-5-fluoronicotinic acid, 96%, Thermo Scientific™
CAS: 82671-06-5 Molecular Formula: C6H2Cl2FNO2 Molecular Weight (g/mol): 209.985 MDL Number: MFCD00799517 InChI Key: LTDGKGCHRNNCAC-UHFFFAOYSA-N Synonym: 2,6-dichloro-5-fluoronicotinic acid,2,6-dichloro-5-fluoro-nicotinic acid,2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid,2,6-dichloro-5-fluoro-pyridine-3-carboxylic acid,2,6-dichloro-5-fluoro nicotinic acid,3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro,pubchem10943,acmc-1bluq,ksc448a5j,2,6-dichloro-5-fluoro-nicotinicacid PubChem CID: 2733659 IUPAC Name: 2,6-dichloro-5-fluoropyridine-3-carboxylic acid SMILES: C1=C(C(=NC(=C1F)Cl)Cl)C(=O)O