Learn More
2,6-Dichloro-4-(trifluoromethyl)pyridin-3-amine, 97%, Thermo Scientific™
Product Code. 10205624
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10205624 25 g Amber Glass Bottle 25g
10715283 1 g Amber Glass Bottle 1g
10149032 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10205624 Supplier Thermo Scientific Maybridge Supplier No. SPB06360EB
 Restricted Item
Estimated Shipment: 20-11-2026
Chemical Identifiers
CAS 175277-67-5
Molecular Formula C6H3Cl2F3N2
Molecular Weight (g/mol) 231.00
MDL Number MFCD00173957
InChI Key BVJJNBRZQVLPIC-UHFFFAOYSA-N
PubChem CID 2735898
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)pyridin-3-amine
SMILES NC1=C(Cl)N=C(Cl)C=C1C(F)(F)F
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Synonym 2,6-dichloro-4-trifluoromethyl pyridin-3-amine, 3-amino-2,6-dichloro-4-trifluoromethyl pyridine, 3-amino-2,6-dichloro-4-trifluoromethyl-pyridine, pubchem2205, 2,6-dichloro-3-amino-4-trifluoromethylpyridine, 2,6-dichloro-4-trifluoromethyl-3-pyridinamine, 2,6-dichloro-4-trifluoromethyl-3-pyridylamine, 2,6-dichloro-4-trifluoromethyl-pyridin-3-ylamine, 3-pyridinamine,2,6-dichloro-4-trifluoromethyl, 3-pyridinamine, 2,6-dichloro-4-trifluoromethyl
Formula Weight 231
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)pyridin-3-amine