accessibility menu, dialog, popup

UserName

CAS RN 175277-67-5

NClClNH2FFF

CAS RN 175277-67-5

IUPAC Name: 2,6-dichloro-4-(trifluoromethyl)pyridin-3-amine
Synonyms: 2,6-dichloro-4-(trifluoromethyl)pyridin-3-amine
Molecular Weight (g/mol): 231
Molecular Formula: C6H3Cl2F3N2
InChi Key: BVJJNBRZQVLPIC-UHFFFAOYSA-N
SMILES: NC1=C(Cl)N=C(Cl)C=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results